17α-Hydroxyprogesterone ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:68-96-2
:
Formula:C₂₁H₃₀O₃
MDL Number:MFCD00003659
:330.46 g/mol
Merck Index:14,04839
Melting Pt:220 °C
EINECS:200-699-4
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Melting point:217.0 to 221.0 deg-C
- Specific rotation [a]20/D:+85.0 to +95.0 deg (C=1, CHCl3)