Order Entry
Sweden
ContactUsLinkComponent
17α-Hydroxyprogesterone
17α-Hydroxyprogesterone
Catalog # EHERC14241000
Supplier:  EHRENSTORFER
CAS Number:  
17α-Hydroxyprogesterone
Catalog # EHERC14241000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,1 g
  • Seq:
    3020

Specifications

About this item

 :
 :330.46 g/mol
EINECS:200-699-4
CAS Number:68-96-2
Formula:C₂₁H₃₀O₃
MDL Number:MFCD00003659