Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Diethyl (3-aminopropyl)phosphonate
Diethyl (3-aminopropyl)phosphonate 95%
Diethyl (3-aminopropyl)phosphonate 95%
Corrosive
Diethyl (3-aminopropyl)phosphonate 95%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Corrosive
Diethyl (3-aminopropyl)phosphonate 95%
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
4402-24-8
:
Formula:
C₇H₁₈NO₃P
MDL Number:
MFCD11110596
:
195.20 g/mol
Density:
1.053
EINECS:
224-535-6
ADR:
8,III
:
GHS05
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAH51874
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation