Triethyl-2-phosphonopropionate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3699-66-9
:
Formula:C₉H₁₉O₅P
MDL Number:MFCD00009159
Boiling Pt:143…144 °C (12 mmHg)
Density:1.111
Flash Pt:88 °C (190 °F)
:238.22 g/mol
Storage Temperature:Ambient
EINECS:223-033-4