Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Nitrocinnamaldehyde
2-Nitrocinnamaldehyde ≥98.0% (by GC)
2-Nitrocinnamaldehyde ≥98.0% (by GC)
:
TCI
2-Nitrocinnamaldehyde ≥98.0% (by GC)
:
TCI
:
1466-88-2
:
:
C₉H₇NO₃
:
MFCD00007188
:
177.16 g/mol
:
215-988-0
:
GHS07
Appearance:
Light yellow to Brown to Dark green powder to crystal
Purity (GC):
min. 98.0 %
Melting point:
126.0 to 130.0 deg-C
Manual Bioscience and Analyticalscience