Order Entry
Sweden
ContactUsLinkComponent
Ponceau S ≥85.0% (by total nitrogen basis)
Ponceau S ≥85.0% (by total nitrogen basis)
Catalog # TCIAA1600-25G
Supplier:  TCI
CAS Number:  
undefined
Ponceau S ≥85.0% (by total nitrogen basis)
Catalog # TCIAA1600-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Orange to Brown to Dark red powder to crystal
  • Purity (with Total Nitrogen):
    min. 85.0 %
  • Lambda max.:
    515.0 to 525.0 nm (H2O)
  • Absorbance (E1%1cm):
    min. 470 (H2O, 515.0 to 525.0 nm)
  • Drying loss:
    max. 15.0 %

Specification Test Results

About this item

 :
 :760.56 g/mol
EINECS:228-319-2
 :GHS07
Storage Temperature:Ambient
CAS Number:6226-79-5
Formula:C₂₂H₁₂N₄Na₄O₁₃S₄
MDL Number:MFCD00003892