
2,2-Dimethylglutaric anhydride ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:2938-48-9
:
Formula:C₇H₁₀O₃
MDL Number:MFCD00006682
:142.1525 g/mol
Boiling Pt:270°C
EINECS:220-922-9
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Morpholine Method):min. 98.0 %
- Melting point:38.0 to 42.0 deg-C
- Solubility in Toluene:almost transparency