β-Cyclodextrine ≥99.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:7585-39-9
:
Formula:C₄₂H₇₀O₃₅
MDL Number:MFCD00078139
:1134.98 g/mol
Merck Index:14,02718
EINECS:231-493-2
Specification Test Results
- Appearance:White powder to crystaline
- Purity (HPLC):min. 99.0 area%
- Specific rotation [a]20/D:+161.0 to +165.0 deg (C=1, Water) (calcd.on anh.substance)
- Solubility in hot Water:almost transparency
- Water:max. 5.0 %