

Isobutylene oxide (2,2-dimethyloxirane, 1,2-epoxy-2-methylpropane) ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:558-30-5
:
Formula:C₄H₈O
MDL Number:MFCD00066354
:72.11 g/mol
Boiling Pt:52°C
Flash Pt:–32°C
EINECS:209-193-8
UN:1993
ADR:3,II
:GHS02
:GHS07
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %