
Fumaric acid monoisopropyl ester ≥97.0% (by GC, titration analysis)
Catalog # TCIAM1784-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 97.0 %
- Purity (Neutralization titration):min. 97.0 %
- Melting point:47.0 to 51.0 deg-C
Specification Test Results
About this item
:
:158.15 g/mol
Melting Pt:48°C
:GHS07
Storage Temperature:Refrigerator
CAS Number:7529-87-5
Formula:C₇H₁₀O₄