
1,2,3,4-Tetrahydronaphthalen-1-one (α-tetralone) ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:529-34-0
:
Formula:C₁₀H₁₀O
MDL Number:MFCD00001688
:146.19 g/mol
Boiling Pt:257°C
Flash Pt:129°C
EINECS:208-460-6
:GHS07
Specification Test Results
- Appearance:Colorless to Yellow to Green clear liquid
- Purity (GC):min. 98.0 %