
(±)-α-Terpineol ≥95.0% (by GC)
Leverantör: TCI
Certifikat
Om denna produkt
CAS-nummer:98-55-5
:
Formel:C₁₀H₁₈O
MDL-nummer:MFCD00001557
:154.25 g/mol
Merck Index:14,09171
Kokande Pt:218°C
EINECS:202-680-6
:GHS07
Specifikationstestresultat
- Appearance:White or Colorless to Light yellow powder to lump to clear liquid
- Purity (GC):min. 95.0 %