Cyclopentanone ≥99.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:120-92-3
:
Formula:C₅H₈O
MDL Number:MFCD00001409
:84.12 g/mol
Merck Index:14,02743
Boiling Pt:130°C
Melting Pt:–51°C
Density:0,941 g/cm³ (20 °C)
Flash Pt:34°C
Storage Temperature:Ambient
EINECS:204-435-9
UN:2245
ADR:3,III
:GHS02
:GHS07
Specification Test Results
- Appearance:Colorless clear liquid
- Purity (GC):min. 99.0 %