2-Chloro-6-nitrotoluene ≥99.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:83-42-1
:
Formula:C₇H₆ClNO₂
MDL Number:MFCD00007205
:171.58 g/mol
Boiling Pt:241°C
Melting Pt:35…37 °C
Storage Temperature:Ambient
EINECS:201-475-9
UN:3457
ADR:6.1,III
:GHS06
:GHS09
Specification Test Results
- Appearance:White to Orange to Green powder to lump
- Purity (GC):min. 99.0 %