1,4,7,10-Tetraazacyclododecane tetrahydrochloride ≥98.0% (by total nitrogen basis)
Supplier: TCI
Certificates
About this item
CAS Number:10045-25-7
:
Formula:C₈H₂₄Cl₄N₄
MDL Number:MFCD00068484
:318.12 g/mol
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (with Total Nitrogen):min. 98.0 %