Allylpentafluorobenzene ≥98.0% (by GC)
Leverantör: TCI
Certifikat
Om denna produkt
CAS-nummer:1736-60-3
:
Formel:(C₆F₅)CH₂CHCH₂
MDL-nummer:MFCD00000301
:208.13 g/mol
Kokande Pt:148 - 149 °C (1013 hPa)
Densitet:1,35 g/cm³ (20 °C)
Flash Pt:42°C
EINECS:217-083-6
FN:1993
ADR:3,III
:GHS02
:GHS07
Specifikationstestresultat
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 98.0 %