1-Methyl-2-phenylindole ≥99.0% (by total nitrogen basis)
Supplier: TCI
Certificates
About this item
CAS Number:3558-24-5
:
Formula:C₁₅H₁₃N
MDL Number:MFCD00022892
:207.27 g/mol
EINECS:222-618-1
Specification Test Results
- Purity (with Total Nitrogen):min. 99.0 %
- Melting point:98.0 to 102.0 deg-C