Mycophenolic acid ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAM2216-1G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:141.0 to 145.0 deg-C
Specification Test Results
About this item
:
:320.34 g/mol
Merck Index:14,06327
EINECS:246-119-3
:GHS07
Storage Temperature:Refrigerator
CAS Number:24280-93-1
Formula:C₁₇H₂₀O₆
MDL Number:MFCD00036814