Order Entry
Sweden
ContactUsLinkComponent
Aniline ≥98.0% (by GC, titration analysis)
Corrosive
Toxic
Health
Environment
Aniline ≥98.0% (by GC, titration analysis)
Catalog # TCIAA0463-500G
Supplier:  TCI
CAS Number:  
undefined
Corrosive
Toxic
Health
Environment
Aniline ≥98.0% (by GC, titration analysis)
Catalog # TCIAA0463-500G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light orange to Yellow clear liquid
  • Purity (GC):
    min. 98.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %

Specification Test Results

About this item

 :
 :93.13 g/mol
Merck Index:14,00659
Boiling Pt:184°C
Melting Pt:-6°C
Melting Pt:–6°C
Density:1,019 g/cm³ (20 °C)
Flash Pt:75°C
Storage Temperature:Ambient
EINECS:200-539-3
UN:1547
ADR:6.1,II
 :GHS05
 :GHS06
 :GHS08
 :GHS09
CAS Number:62-53-3
Formula:C₆H₅NH₂
MDL Number:MFCD00007629