Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,3,5,6-Tetrafluorobenzaldehyde
2,3,5,6-Tetrafluorobenzaldehyde ≥94.0% (by GC)
2,3,5,6-Tetrafluorobenzaldehyde ≥94.0% (by GC)
:
TCI
2,3,5,6-Tetrafluorobenzaldehyde ≥94.0% (by GC)
:
TCI
:
19842-76-3
:
:
C₇H₂F₄O
:
MFCD00167387
:
178.09 g/mol
:
60°C
:
74°C
:
GHS07
Appearance:
Colorless to Light orange to Yellow clear liquid
Purity (GC):
min. 94.0 %