Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Bromophenyl methyl sulfone
3-Bromophenyl methyl sulphone ≥98.0% (by GC)
3-Bromophenyl methyl sulphone ≥98.0% (by GC)
:
TCI
3-Bromophenyl methyl sulphone ≥98.0% (by GC)
:
TCI
:
34896-80-5
:
MFCD21364720
:
C₇H₇BrO₂S
:
MFCD03095326
:
235.1 g/mol
:
GHS07
Appearance:
White to Light yellow powder to crystal
Purity (GC):
min. 98.0 %
Melting point:
103.0 to 107.0 deg-C