

trans-3-Octene ≥98.0% (by GC)
TCIAO0134-5ML
: TCI
:
- Pack type:Glass bottle
- Pk:5 mL
- Seq:2210
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
:
:112.22 g/mol
:123°C
:–110 °C
:0,7152 g/cm³ (20 °C)
:14°C
:Ambient
:238-990-3
:3295
:3,II
:GHS02
:GHS08
:14919-01-8
:C₈H₁₆
:MFCD00039971