Order Entry
Sweden
ContactUsLinkComponent
1-Octene ≥99.0% (by GC)
1-Octene ≥99.0% (by GC)
Catalog # TCIAO0251-25ML
Supplier:  TCI
CAS Number:  
undefined
1-Octene ≥99.0% (by GC)
Catalog # TCIAO0251-25ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 mL
  • Seq:
    2315

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 99.0 %

Specification Test Results

About this item

 :
 :112.22 g/mol
Merck Index:14,01764
Boiling Pt:123°C
Melting Pt:–102 °C
Density:0,7152 g/cm³ (20 °C)
Flash Pt:10°C
EINECS:203-893-7
UN:3295
ADR:3,II
 :GHS02
 :GHS08
 :GHS09
Storage Temperature:Ambient
CAS Number:111-66-0
Formula:C₈H₁₆
MDL Number:MFCD00009548