Order Entry
Sweden
ContactUsLinkComponent
 
 
1-Hexadecene ≥90.0% (by GC)
 :  TCI
 
undefined
 
1-Hexadecene ≥90.0% (by GC)
 :  TCI

 

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 90.0 %