Order Entry
Sweden
ContactUsLinkComponent
2-Methylheptane ≥98.0% (by GC)
 
 
 
 
2-Methylheptane ≥98.0% (by GC)
  TCIAM0519-5G
 :  TCI
 :  
undefined
 
 
 
 
2-Methylheptane ≥98.0% (by GC)
  TCIAM0519-5G
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

 

 

  • Appearance:
    Colorless clear liquid
  • Purity (GC):
    min. 98.0 %

 

 

 :
 :114.23 g/mol
 :117°C
 :–109 °C
 :0,698 g/cm³ (20 °C)
 :4°C
 :Ambient
 :209-747-9
 :1262
 :3,II
 :GHS02
 :GHS07
 :GHS08
 :GHS09
 :592-27-8
 :250-610-8
 :C₈H₁₈
 :MFCD00009518