1-Octadecanol ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:112-92-5
:
Formula:H₃C(CH₂)₁₆CH₂OH
MDL Number:MFCD00002823
:270.5 g/mol
Merck Index:14,08805
Boiling Pt:210°C
Melting Pt:56…57 °C
Density:0,805 to 0.815 g/cm³ (60 °C)
Storage Temperature:Ambient
EINECS:204-017-6
Specification Test Results
- Appearance:White pellet
- Purity (GC):min. 98.0 %
- Melting point:57.0 to 60.0 deg-C
- Solubility in hot Methanol:almost transparency