Order Entry
Sweden
ContactUsLinkComponent
Behenic acid ≥80.0% (by GC)
Behenic acid ≥80.0% (by GC)
Supplier:  TCI
Certificates
undefined
Behenic acid ≥80.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystaline
  • Purity (GC):
    min. 80.0 %
  • Melting point:
    76.0 to 81.0 deg-C

Specification Test Results

Related Information