

3-Pentanon ≥98.0%
Supplier: TCI
Certificates
About this item
CAS Number:96-22-0
:
Formula:C₅H₁₀O
MDL Number:MFCD00009320
:86.13 g/mol
Merck Index:14,03121
Boiling Pt:102°C
Melting Pt:–40 °C
Density:0,8148 g/cm³ (20 °C)
Flash Pt:13°C
Storage Temperature:Omgivande
EINECS:202-490-3
UN:1156
ADR:3,II
:GHS02
:GHS07
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %