0,00 kr
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Sweden
ContactUsLinkComponent
7-Methoxyflavone
7-Methoxyflavone ≥98.0% (by GC)
7-Methoxyflavone ≥98.0% (by GC)
:
TCI
7-Methoxyflavone ≥98.0% (by GC)
:
TCI
:
22395-22-8
:
:
C₁₆H₁₂O₃
:
MFCD00017462
:
252.27 g/mol
Appearance:
White to Light yellow powder to crystal
Purity (GC):
min. 98.0 %
Melting point:
109.0 to 113.0 deg-C
Manual Bioscience and Analyticalscience