Order Entry
Sweden
ContactUsLinkComponent
(±)-Epichlorhydrin ≥99.0% (by GC)
Flammable
Corrosive
Toxic
Health
(±)-Epichlorhydrin ≥99.0% (by GC)
Supplier:  TCI
Certificates
undefined
Flammable
Corrosive
Toxic
Health
(±)-Epichlorhydrin ≥99.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless clear liquid
  • Purity (GC):
    min. 99.0 %
  • Solubility in EtOH (95vol%):
    almost transparency

Specification Test Results

Related Information

Frequently Bought Together