Methyl violet 2B
Supplier: TCI
About this item
Synonyms: Basic Violet 1, 4-[4,4'-Bis(dimethylamino) benzhydrylidene]cyclohexa-2,5-dien-1-ylidene]methylammonium chloride
- CAS Number:8004-87-3
- Formula:C₂₄H₂₈ClN₃
- MDL Number:MFCD00012568
- Molecular Weight:393.96 g/mol
- Melting Pt:137 °C
- Storage Temperature:Ambient
- ADR:9,III
2 Options Available Below
Product Details & Documents
Synonyms: Basic Violet 1, 4-[4,4'-Bis(dimethylamino) benzhydrylidene]cyclohexa-2,5-dien-1-ylidene]methylammonium chloride
- CAS Number:8004-87-3
- Formula:C₂₄H₂₈ClN₃
- MDL Number:MFCD00012568
- Molecular Weight:393.96 g/mol
- Melting Pt:137 °C
- Storage Temperature:Ambient
- ADR:9,III
Documents
Specifications for Product Family
Specification Test Results
- Appearance:Green to Dark green powder to crystal
- Lambda max.:583.0 to 587.0 nm (H2O)
- Absorbance:min. 0.320 (2mg/L, H2O, 583.0 to 587.0 nm) (calcd.on dried substance)
- abs-ratio (a/b):1.05 to 1.20 (E570/E600)
- Drying loss:max. 10.0 %



