Orange G
Glass bottle, 25 g
Katalog # TCIAO0093-25G
Leverantör: TCI
Gå till:
Product Details & Documents
Synonymer: Disodium 7-hydroxy-8-phenylazonaphthalene-1,3-disulphonate, Wool Orange 2G, Orange G sodium salt, CI 16230, Acid Orange 10
- CAS-nummer:1936-15-8
- Formel:C₁₆H₁₀N₂Na₂O₇S₂
- MDL-nummer:MFCD00012457
- Molecular Weight:452.38 g/mol
- Smält Pt:141 °C
- Densitet:0,45 to 0.47 g/cm³ (20 °C)
- Förvaringstemperatur:Ambient
- EINECS:217-705-6
Specifikationer
Specifikationstestresultat
- Appearance:Light yellow to Yellow to Orange powder to crystal
- Lambda max.:476.0 to 482.0 nm (H2O)
- Absorbance:min. 0.400 (10mg/L, H2O, 476.0 to 482.0 nm) (calcd.on dried substance)
- Drying loss:max. 5.0 %
Specifikationer
- Pack type:Glass bottle
- Pk:25 g
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



