
Neocuproin hemihydrat ≥98.0% (by HPLC, titration analysis)
TCIAD0771-1G
: TCI
:
- Förpackningstyp:Glass bottle
- Förp.:1 g
- Seq:3047
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 % (calcd.on anh.substance)
- Melting point:160.0 to 164.0 deg-C
- Solubility in Methanol:almost transparency
:
:434.53 g/mol
:14,06449
:207-601-9
:GHS07
:34302-69-7
:C₂₈H₂₆N₄O
:MFCD00149306