
Methyl-4-chloro-3-nitrobenzoate ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:14719-83-6
:
Formula:C₈H₆ClNO₄
MDL Number:MFCD00016989
:215.59 g/mol
:GHS07
Specification Test Results
- Appearance:White to Light yellow to Green powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:79.0 to 83.0 deg-C