Methyl red ≥92.0% (by HPLC)
Catalog # TCIAM0702-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:Orange to Brown to Dark purple powder to crystal
- Purity (HPLC):min. 92.0 area%
- Absorbance:min. 0.30 (5mg/L, pH 6.6 buffer sol., 425.0 to 435.0nm)
- Drying loss:max. 5.0 %
- Transition interval (pH):(purplish red)4.2 to 6.2 (yellow)
Specification Test Results
About this item
:
Merck Index:14,06119
Melting Pt:178…182 °C
Density:1,31 g/cm³ (20 °C)
:269.3 g/mol
Storage Temperature:Ambient
EINECS:207-776-1
CAS Number:493-52-7
Formula:C₁₅H₁₅N₃O₂
MDL Number:MFCD00002425