D-α-Phenylalanine ≥98.0% (by HPLC, titration analysis)
: TCI
:673-06-3
:
:C₉H₁₁NO₂
:MFCD00004270
:165.19 g/mol
:14,07271
:295 °C (1013 hPa)
:211-603-5
- Appearance:White powder to crystal
- Purity (HPLC):min. 98.0 area%
- Optical purity (LC):min. 98.0 ee%
- Purity (Nonaqueous Titration):min. 98.0 %
- Solubility in hot Water:almost transparency