Order Entry
Sweden
ContactUsLinkComponent
N,O-Bis(trimethylsilyl)trifluoroacetamide ≥95.0% (by GC)
Flammable
Corrosive
N,O-Bis(trimethylsilyl)trifluoroacetamide ≥95.0% (by GC)
Supplier:  TCI
Certificates
undefined
Flammable
Corrosive
N,O-Bis(trimethylsilyl)trifluoroacetamide ≥95.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Purity (GC):
    min. 95.0 %

Specification Test Results

Related Information

Frequently Bought Together