
Tetramethylammonium fluoride tetrahydrate ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:17787-40-5
:
Formula:C₄H₁₂FN·4H₂O
MDL Number:MFCD00149968
:165.21 g/mol
Melting Pt:39…44 °C
Density:1,115 g/cm³ (20 °C)
Storage Temperature:Refrigerator
EINECS:206-769-0
ADR:6.1,III
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to lump
- Purity (Nonaqueous Titration):min. 98.0 %
- Water:40.0 to 45.0 %
- TMAF-HF:max. 2.0 %