Tetrapropylammonium iodide ≥98.0% (by titrimetric analysis)
Leverantör: TCI
Certifikat
Om denna produkt
CAS-nummer:631-40-3
:
Formel:C₁₂H₂₈IN
MDL-nummer:MFCD00011842
:313.27 g/mol
Smält Pt:283 °C
Densitet:1,317 g/cm³ (20 °C)
EINECS:211-157-1
:GHS07
Specifikationstestresultat
- Appearance:White to Almost white powder to crystal
- Purity (Nonaqueous Titration):min. 98.0 %
- Solubility in Water:almost transparency