Order Entry
Sweden
ContactUsLinkComponent
 
Tributylphosphine oxide ≥95.0%
  TCIAT0921-25G
 :  TCI
 :  
undefined
Tributylphosphine oxide ≥95.0%
  TCIAT0921-25G
 :  TCI
 :  

 

  • Förpackningstyp:
    Glass bottle
  • Förp.:
    25 g
  • Seq:
    3308

 

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 95.0 %

 

 

 :
 :218.32 g/mol
 :348 °C (1013 hPa)
 :62…63 °C
 :212-394-3
 :1759
 :8,III
 :GHS05
 :GHS07
 :814-29-9
 :C₁₂H₂₇OP
 :MFCD00002082