Order Entry
Sweden
ContactUsLinkComponent
4-Chloro-2-methoxyphenol ≥98.0% (by GC)
4-Chloro-2-methoxyphenol ≥98.0% (by GC)
Catalog # TCIAC1517-5G
Supplier:  TCI
CAS Number:  
undefined
4-Chloro-2-methoxyphenol ≥98.0% (by GC)
Catalog # TCIAC1517-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White or Colorless to Yellow to Green powder to lump to clear liquid
  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
 :158.58 g/mol
Boiling Pt:241°C
Density:1,304 g/cm³ (20 °C)
 :GHS07
CAS Number:16766-30-6
Formula:C₇H₇ClO₂
MDL Number:MFCD00070774