

Hexamethyldisiloxane ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:107-46-0
:
Formula:C₆H₁₈OSi₂
MDL Number:MFCD00008265
:162.38 g/mol
Boiling Pt:100°C
Melting Pt:–59 °C
Density:0,764 g/cm³ (20 °C)
Flash Pt:–2°C (closed cup)
Storage Temperature:Ambient
EINECS:203-492-7
UN:1993
ADR:3,II
REACH:01-2119496108-31
:GHS02
:GHS07
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %