1-Methylpyrrole ≥99.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:96-54-8
:
Formula:C₅H₇N
MDL Number:MFCD00005345
:81.12 g/mol
Boiling Pt:113°C
Melting Pt:–57 °C
Density:0,91 g/cm³ (20 °C)
Flash Pt:16°C
Storage Temperature:Ambient
EINECS:202-513-7
UN:1993
ADR:3,II
:GHS02
:GHS07
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 99.0 %