Order Entry
Sweden
ContactUsLinkComponent
Benzyl nicotinate ≥98.0% (by GC)
Benzyl nicotinate ≥98.0% (by GC)
Catalog # TCIAN0083-25G
Leverantör:  TCI
CAS-nummer:  
undefined
Benzyl nicotinate ≥98.0% (by GC)
Catalog # TCIAN0083-25G
Leverantör:  TCI
CAS-nummer:  

Specifikationer

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifikationer

Specifikationstestresultat

  • Appearance:
    White to Light yellow to Light orange powder to lump
  • Purity (GC):
    min. 98.0 %

Specifikationstestresultat

Om denna produkt

 :
 :213.24 g/mol
Merck Index:14,06526
Kokande Pt:189°C
Smält Pt:24 °C
Densitet:1,17 g/cm³ (20 °C)
EINECS:202-332-3
 :GHS07
CAS-nummer:94-44-0
Formel:C₁₃H₁₁NO₂
MDL-nummer:MFCD00023584