1,4-Diazabicyclo[2.2.2]octane ≥98.0% (by GC, titration analysis)
: TCI
: DABCO
:280-57-9
:
:C₆H₁₂N₂
:MFCD00006689
:112.17 g/mol
:14,09669
:174 °C (1013 hPa)
:155…158 °C
:1,14 g/cm³ (20 °C)
:62 °C
:Ambient
:205-999-9
:3263
:8,III
:GHS05
:GHS07
- Environmentally Preferable:
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:155.0 to 161.0 deg-C
- Solubility in Water:almost transparency