Benzyl salicylate ≥99.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:118-58-1
:
Formula:C₁₄H₁₂O₃
MDL Number:MFCD00020034
:228.25 g/mol
Merck Index:14,01144
Boiling Pt:194°C
Melting Pt:23.9 °C
Density:1,1755 g/cm³ (20 °C)
Flash Pt:184°C
EINECS:204-262-9
:GHS07
Specification Test Results
- Appearance:White or Colorless to Light yellow powder to lump to clear liquid
- Purity (GC):min. 99.0 %