4-Methoxybenzyl alcohol ≥98.0% (by GC)
Leverantör: TCI
Certifikat
Om denna produkt
CAS-nummer:105-13-5
:
Formel:C₈H₁₀O₂
MDL-nummer:MFCD00004653
:138.17 g/mol
Merck Index:14,00665
Kokande Pt:259°C
Smält Pt:20…24 °C
Densitet:1,1 g/cm³ (20 °C)
Flash Pt:148°C
EINECS:203-273-6
:GHS07
Förvaringstemperatur:Ambient
Specifikationstestresultat
- Appearance:White or Colorless to Almost white or Almost colorless powder to lump to clear liquid
- Purity (GC):min. 98.0 %