Order Entry
Sweden
ContactUsLinkComponent
 
3'-Nitropropiophenone ≥98.0% (by GC)
 :  TCI
 
undefined
3'-Nitropropiophenone ≥98.0% (by GC)
 :  TCI

 

  • Appearance:
    White to Yellow to Green powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    98.0 to 102.0 deg-C