3,4,7,8-Tetramethyl-1,10-phenanthroline ≥98.0% (by HPLC, titration analysis)
TCIAT0847-1G
: TCI
:
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Solubility in hot Methanol:almost transparency
:
:236.32 g/mol
:386 °C (1013 hPa)
:284 °C
:216-762-4
:1660-93-1
:C₁₆H₁₆N₂
:MFCD00004974