
(R)-(-)-2-Amino-1-propanol ≥98.0% (by GC, titration analysis)
: TCI
:35320-23-1
:
:C₃H₉NO
:MFCD00064413
:75.11 g/mol
:173 °C (1013 hPa)
:0,96 g/cm³ (20 °C)
:84°C
:Refrigerator
:2735
:8,II
:GHS05
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 98.0 %
- Optical purity (GC):min. 98.0 ee%
- Purity (Nonaqueous Titration):min. 98.0 %
- Specific rotation [a]20/D:-20.0 to -24.0 deg (C=2, EtOH)